Stampa…
VWR®, Side Access Rack for Cryoboxes, Height 50 mm
Fornitore: VWR Collection
479-2265EA 97.7 EUR
479-2265 479-2266 479-2267 479-2268 479-2269 479-2270 479-2271
VWR®, Side Access Rack for Cryoboxes, Height 50 mm
Rack Supporti per cryobox
I rack per congelatori come sistemi di stoccaggio e inventario per congelatori ULT verticali facilitano l'organizzazione dei campioni.
  • Different height and depth dimensions are available and can be used in all freezer brands
  • Racks for standard 2" cryoboxes (133×133×51 mm)
  • Made from corrosion resistant stainless steel with smoothed edges
  • Organise and find your samples quickly
These racks are made from corrosion resistant stainless steel with smoothed edges. Side access racks are a cost-effective freezer rack that will fit into any freezer.