Stampa…

Manganese(II) fluoride ≥99.99% (metals basis), Puratronic®

Fornitore: Thermo Fisher Scientific
Pericolo

47109.14 47109.22
47109.14EA 238 EUR
47109.14 47109.22
Manganese(II) fluoride ≥99.99% (metals basis), Puratronic®
Manganese(II) fluoride

Purity: 99.99% CAS No.: 7782-64-1 Molecular Formula: F2Mn Formula Weight: 92.93 Physical Form: Crystalline Powder Density: 3.98 Sensitivity: Hygroscopic Appearance: Red Melting Point: 856° Solubility: Insoluble in water, alcohol. Soluble in dilute HF, conc. HCl or HNO3 UN No.: UN3288 MDL No.: MFCD00016222

Formula: F₂Mn
Punto di fusione: 856 °C
Densità: 3,98
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016222
CAS Number: 7782-64-1
EINECS: 231-960-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,5729

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR