Stampa…

n-Esano, anhydrous, molecular sieves_over

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Hexyl hydride, Hexanen, Hexane

Argon_Chemseal

47104.AK 47104.K2
47104.AKEA 54.3 EUR
47104.AK 47104.K2
n-Esano, anhydrous, molecular sieves_over
n-Esano

CAS No.: 110-54-3 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -23°(-9°F) Physical Form: Liquid Density: 0.659 Refractive Index: 1.3750 Appearance: Colourless Melting Point: -95° Boiling Point: 69° Beilstein Reference: 1730733 UN No.: UN1208 MDL No.: MFCD00009520

Formula: H₃C(CH₂)₄CH₃
Peso molecolare: 86.18 g/mol
Punto di ebollizione: 69 °C
Punto di fusione: –95 °C
Densità: 0.659
Flash Pt: –23 °C (–9 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 14,4694
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR