Stampa…
Incubatori refrigerati MIR, PHCbi
Fornitore: PHC Corporation
MIR-154-PE
471-0671EA 5230 EUR
471-0671
Incubatori refrigerati MIR, PHCbi
Incubatori Incubatori refrigerati
These cooled incubators are designed to meet a variety of advanced experimental needs, ranging from microorganism cultures and plant germination tests to various constant temperature experiments. Design includes a window made from triple layer glass (which can be shaded), lighting in the chamber (15 Watt), cable pass through as standard. MIR-154/MIR-254 only: Control panel with LCD display. DC fan to protect medium from drying out. Reversible door for left or right-hand door opening. Remote monitoring with DAQ (data acquisition) software. MIR-554: Auto diagnostic function, programmable withMore Product Information