Stampa…

Acido cloridrico concentrate for dilution to 0.1 N in soluzione acquosa, DILUT-IT®, Laboratory Reagent, J.T.Baker®

Fornitore: Avantor
Attenzione

4655.EA 28.1 EUR
4655.
Acido cloridrico concentrate for dilution to 0.1 N in soluzione acquosa, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Acido cloridrico

(1/10 equiv.=3.646 g HCl)

Formula: HCl
Densità: 1 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

For Laboratory,Research,or Manufacturing Use Passes Test
Normality (by titrimetry) Passes Test
(Makes 0.1N solution after dilution to 1000 mL)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR