Stampa…

Magnesio tiosolfato esaidrato ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Magnesium hyposulphite hexahydrate

46114.22 46114.36
46114.22EA 29 EUR
46114.22 46114.36
Magnesio tiosolfato esaidrato ≥99%
Magnesio tiosolfato esaidrato

Purity: 99% CAS No.: 13446-30-5 Molecular Formula: MgO3S2.6H2O Formula Weight: 244.53 (136.44anhy) Physical Form: Crystalline Density: 1.818 Appearance: White Melting Point: 170° (-3H2O) Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,5694 MDL No.: MFCD00169871

Formula: MgO₃S₂·6H₂O
Peso molecolare: 244.53 g/mol
Punto di fusione: 170 °C (-3H₂O)
Densità: 1.818
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00169871
CAS Number: 13446-30-5
EINECS: 233-340-5
Merck Index: 13,05718

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR