Stampa…
Optical windows
Fornitore: Thermo Fisher Scientific
45387.KS 45386.KS 45560.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
45387.KSEA 276 EUR
45387.KS 45386.KS 45560.KS 45666.KS 45813.KS 45856.KS 45925.KS 45986.KS 46057.KS 46089.KS
Optical windows
Finestre ottiche
These optical windows are ideally stored at ambient temperatures.
  • Clear aperture 85%
  • Surface finish 60/40 or 40/20