Stampa…

Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®

Fornitore: Thermo Fisher Scientific

45290.AK
45290.AKEA 145 EUR
45290.AK
Palladium nitrate and magnesium nitrate, matrix modifier solution, Specpure®
Standard Standard cationici Standard multi-elemento

Pre-mixed matrix modifier are designed for use with graphite furnace atomic absorption (GFAA).


UN No.: UN3264
MDL No.: MFCD09910543

Visualizza i codici

Specifiche

Environmentally Preferable
N. MDL MFCD09910543

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR