Stampa…
Cartridge burner, Teclu
Fornitore: USBECK Laborgeräte
1421
451-0177EA 62.6 EUR
451-0177
Cartridge burner, Teclu
Termostati Bruciatori a gas
Cartridge burner with air regulation. Maximum temperature of 1640 °C.