Stampa…

Vinylmagnesium bromide 1 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®

Fornitore: Thermo Fisher Scientific
Pericolo

445971000 445978000
445971000.EA 111 EUR
445971000. 445978000.
Vinylmagnesium bromide 1 M in 2-MeTHF (2-methyltetrahydrofuran), AcroSeal®
Vinylmagnesium bromide
Formula: C₂H₃BrMg
Densità: 0,965 g/cm³
Flash Pt: –11 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000042
CAS Number: 1826-67-1
UN: 3399
ADR: 4.3,I

Visualizza i codici

Risultati del test di specificazione

Appearance Yellow to red to brown Solution (May develop some turbidity or precipitate)
Titration Active base 0.80 to 1.10 M


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR