Stampa…
VWR® Mini Shakers, 230 V
Fornitore: VWR Collection
444-1418EA 1240 EUR
444-1418 444-1422
VWR® Mini Shakers, 230 V
Agitatori Agitatori orbitali
VWR® mini shakers feature variable-speed, microprocessor control, providing reliable, uniform mixing action. Ideal for cell cultures, solubility studies, extraction procedures, diagnostic tests, general mixing, hybridisation, genetics research, bacterial suspensions, staining/destaining, and washing procedures.
  • Choice of models with either 3 or 20 mm orbit
  • LCD display for speed and time
  • Includes platform and mat; ready to attach a variety of optional accessories
  • Ideal for use in non-condensing incubators, CO₂ incubators, or cold rooms
  • RS232 interface