Stampa…

Piombo (II) ioduro, ultra dry ≥99.999% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo

44314.06 44314.14 44314.22
44314.06EA 112 EUR
44314.06 44314.14 44314.22
Piombo (II) ioduro, ultra dry ≥99.999% (metals basis)
Piombo (II) ioduro

Purity: 99.999% CAS No.: 10101-63-0 Molecular Formula: I2Pb Formula Weight: 461.00 Physical Form: -10 Mesh Beads Density: 6.16 Appearance: Yellow Odour: Odourless Melting Point: 402° Boiling Point: 954° Merck Reference: 14,5411 UN No.: 2291 MDL No.: MFCD00011163

Formula: I₂Pb
Punto di ebollizione: 954 °C
Punto di fusione: 402 °C
Densità: 6,16
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011163
CAS Number: 10101-63-0
EINECS: 233-256-9
UN: 2291
ADR: 6.1,III
Merck Index: 14,5411

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR