Stampa…

Nigrosine 10% staining solution, Microscopy

Fornitore: SIGMA ALDRICH MICROSCOPY
Pericolo

Synonyms: CI 50420

MQ100

43925-100ML-F
43925-100ML-FEA 40.6 EUR
43925-100ML-F
Nigrosine 10% staining solution, Microscopy
Nigrosine

Composition: Nigrosin 100 g/l, Formalin 5 ml/l in water


Negative staining method permits visualization of the usually transparent and unstainable capsule of many organisms, most importantly Cryptococcus neoformans.

MDL Number: MFCD00044681
CAS Number: 8005-03-6

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR