Stampa…

Scandium standard dissolved in oil, 1.000 µg/g Sc in mineral oil Specpure® for wear metal analysis

Fornitore: Thermo Fisher Scientific

43129.18
43129.18EA 156 EUR
43129.18
Scandium standard dissolved in oil, 1.000 µg/g Sc in mineral oil Specpure® for wear metal analysis
Standard Standard cationici Standard di scandio

Physical Form: Liquid
MDL No.: MFCD00016323

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR