Stampa…

Palladio (II) cloruro (≥59,5% Pd) ≥99.999% (metals basis) Pd ≥ 59.5%, Premion®

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Palladium dichloride

43085.03 43085.06 43085.14
43085.03EA 199 EUR
43085.03 43085.06 43085.14
Palladio (II) cloruro (≥59,5% Pd) ≥99.999% (metals basis) Pd ≥ 59.5%, Premion®
Palladio (II) cloruro

Purity: 99.999% CAS No.: 7647-10-1 Molecular Formula: PdCl2 Formula Weight: 177.31 Physical Form: Powder Density: 4.0 Appearance: Red-brown Odour: Odourless Melting Point: 675° dec. Merck Reference: 14,6990 MDL No.: MFCD00003558

Formula: PdCl₂
Punto di fusione: 675 °C (dec.)
Densità: 4,0
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003558
CAS Number: 7647-10-1
EINECS: 231-596-2
UN: 3260
ADR: 8,III
Merck Index: 14,6990

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR