Stampa…
Supelco® Organic Reference Standard, Palmitic Acid
Fornitore: Merck
43051-100MG
43051-100MGEA 81.1 EUR
43051-100MG
Supelco® Organic Reference Standard, Palmitic Acid
Standard Standard organici
Certificazioni: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0849d66c5821c63149ccfd0f066925871701b6ea&vid=0849d66c5821c63149ccfd0f066925871701b6ea&prid=web&clid=SAPDAM