Stampa…

Silicon nitride α-phase ≥95%

Fornitore: Thermo Fisher Scientific

42949.30 42949.18 42949.A1
42949.30EA 201 EUR
42949.30 42949.18 42949.A1
Silicon nitride α-phase ≥95%
Silicon nitride

Purity: 95% CAS No.: 12033-89-5 Molecular Formula: Si3N4 85% min (alpha form) Formula Weight: 140.28 Physical Form: 10 Micron Powder, S.A. 5.0m2/g Density: 3.2 Appearance: Pale gray Odour: Odourless Melting Point: 1900° subl. Merck Reference: 14,8497 MDL No.: MFCD00011230

Formula: N₄Si₃
Punto di fusione: 1900 °C (subl.)
Densità: 3,2
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011230
CAS Number: 12033-89-5
EINECS: 234-796-8
Merck Index: 14,8497

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR