Stampa…

Piombo (II) cloruro, ultra dry ≥99.999% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo


Ampuled under argon

42841.04 42841.09 42841.18
42841.04EA 69 EUR
42841.04 42841.09 42841.18
Piombo (II) cloruro, ultra dry ≥99.999% (metals basis)
Piombo (II) cloruro

Purity: 99.999% CAS No.: 7758-95-4 Molecular Formula: PbCl2 Formula Weight: 278.10 Physical Form: Powder, Ampouled under argon Density: 5.85 Melting Point: 501° Boiling Point: 951° Merck Reference: 14,5404 UN No.: 2291 MDL No.: MFCD00011157

Formula: Cl₂Pb
Punto di ebollizione: 951 °C
Punto di fusione: 501 °C
Densità: 5,85
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III
Merck Index: 14,5404

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR