Stampa…

Boron standard dissolved in oil, 5,000 µg/g B in mineral oil Specpure® for wear metal analysis

Fornitore: Thermo Fisher Scientific

42435.18
42435.18EA 101 EUR
42435.18
Boron standard dissolved in oil, 5,000 µg/g B in mineral oil Specpure® for wear metal analysis
Standard Standard cationici Standard di boro

Physical Form: Liquid
MDL No.: MFCD00134034

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR