Stampa…

Acido cloroacetico 99% ACS

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Monochloroacetic acid

41724.22 41724.36 41724.57
41724.22EA 40.4 EUR
41724.22 41724.36 41724.57
Acido cloroacetico 99% ACS
Acido cloroacetico

Purity: 99% CAS No.: 79-11-8 Molecular Formula: C2H3ClO2 Formula Weight: 94.50 Flash Point: 126°(258°F) Physical Form: Crystalline Density: 1.404 Appearance: White Odour: Pungent pH: 1.0 Melting Point: 60-63° Boiling Point: 189° Beilstein Reference: 605438 Merck Reference: 14,2112 UN No.: 1751 MDL No.: MFCD00002683

Formula: ClCH₂COOH
Punto di ebollizione: 189 °C
Punto di fusione: 60…63 °C
Densità: 1,404
Flash Pt: 126 °C (258 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002683
CAS Number: 79-11-8
EINECS: 201-178-4
UN: 1751
ADR: 6.1,II
Merck Index: 14,2112
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2516e0c26c6c26f4f6f3656bc5b4256e7cf08207&vid=9c9b7ad094469d9089eb2f0ce88275d1423c2b46&prid=web&clid=SAPDAM

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR