Stampa…

TCEP HCl (Tris (2-carbossietil) fosfina cloridrato) ≥98%

Fornitore: Thermo Fisher Scientific
Pericolo

40587.02 40587.04 40587.09
40587.02EA 49.6 EUR
40587.02 40587.04 40587.09
TCEP HCl (Tris (2-carbossietil) fosfina cloridrato) ≥98%
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)

Purity: 98% CAS No.: 51805-45-9 Molecular Formula: C9H15O6P.HCl Formula Weight: 286.65 Physical Form: Crystalline powder Sensitivity: Moisture Sensitive Appearance: White Melting Point: 177-178° Solubility: Soluble in water Merck Reference: 14,9085 UN No.: 3261 MDL No.: MFCD00145469


Reducing agent, specific for disulfides

Formula: C₉H₁₆ClO₆P
Punto di fusione: 177…178 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00145469
CAS Number: 51805-45-9
UN: 3261
ADR: 8,II
Merck Index: 14,9085

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR