Stampa…

Igromicina B, Millipore®

Fornitore: Merck Millipore Calbiochem
Pericolo

400051-1 400051-5
400051-1EA 320 EUR
400051-1 400051-5
Igromicina B, Millipore®
Hygromycin B
Formula: C₂₀H₃₇N₃O₁₃
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD06795479
CAS Number: 31282-04-9
EINECS: 250-545-5
UN: 3462
ADR: 6.1,I

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR