Stampa…
Reduced-serum medium, Corning® transfectagro™
Fornitore: Corning
40-300-CV
40-300-CVEA 204.78 EUR
40-300-CV
Reduced-serum medium, Corning® transfectagro™
Terreni per colture cellulari
Corning® transfectagro® Reduced Serum Medium is designed to maximize the growth of a variety of cell types under reduced serum conditions, generally allowing for at least a 50% reduction in serum use with minimal adaptation
Corning® cellgro® transfectaGRO® has also been proven to improve transient transfection efficiency in a variety of cell types. This is a chemically-defined, reduced serum formulation of Eagle’s Minimum Essential Medium with lower levels of calcium and phenol red, and with HEPES for greater buffering capacity.