Stampa…

Silver molybdenum oxide ≥99%

Fornitore: Thermo Fisher Scientific

39650.01 39650.03
39650.01EA 65.5 EUR
39650.01 39650.03
Silver molybdenum oxide ≥99%
Silver molybdenum oxide

Purity: 99% CAS No.: 13765-74-7 Molecular Formula: Ag2MoO4 Formula Weight: 375.68 Physical Form: Powder Density: 6.18 Sensitivity: Light Sensitive Appearance: Beige Odour: Odourless Melting Point: 483° MDL No.: MFCD00053384

Formula: Ag₂MoO₄
Punto di fusione: 483 °C
Densità: 6,18
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00053384
CAS Number: 13765-74-7
EINECS: 237-374-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR