Stampa…

Bis-trispropane, Millipore®

Fornitore: Merck Millipore Calbiochem

394111-100
394111-100EA 346 EUR
394111-100
Bis-trispropane, Millipore®
Bis-trispropane
Formula: C₁₁H₂₆N₂O₆
MDL Number: MFCD00004689
CAS Number: 64431-96-5
EINECS: 264-899-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR