Stampa…
Crucibles, glassy carbon
Fornitore: Thermo Fisher Scientific
39011.KT 38376.KT 39003.KT 39004.KT 39005.KT 39006.KT 39008.KT 39010.KT 39012.KT 39014.KT
39011.KTEA 588 EUR
39011.KT 38376.KT 39003.KT 39004.KT 39005.KT 39006.KT 39008.KT 39010.KT 39012.KT 39014.KT
Crucibles, glassy carbon
Crogioli
These crucibles are manufactured from glassy carbon. Available in conical and cylindrical types.
  • Resistance to all wet decomposition agents
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Glassy carbon, a brittle form of carbon with a randomised structure, has certain specific properties making it appropriate for fields of application outside the scope of carbonMore Product Information