Stampa…
Cooled Incubators, KB PRO Series
Fornitore: Binder
9020-0433 9020-0434 9020-0435 9020-0436 9020-0437 9020-0438
390-1747EA 11753 EUR
390-1747 390-1748 390-1749 390-1750 390-1751 390-1752
Cooled Incubators, KB PRO Series
Incubatori Incubatori refrigerati
The KB PRO series are powerful cooling incubators with energy-efficient cooling with modern inverter cooling unit. The units with proven APT.line™ preheating chamber technology have a temperature range of -20 to +100 °C and comprehensive programming options. The models have a stainless-steel inner chamber and cover a wide range of applications that extend well beyond incubation. Optional light modules are also available to turn the KB PRO into a chamber for ICH photostability testing or plant growth.
  • Cooling with climate-neutral R-290 refrigerant (GWP is 3)
  • Door heating to prevent