Stampa…

Tungsteno 1000 mg/L W in 5% acido nitrico with hydrofluoric acid (max. 1%) (da W) Specpure® Standard monoelemento per ICP

Fornitore: Thermo Fisher Scientific

35770.AD 35770.AE 35770.AP
35770.ADEA 54.9 EUR
35770.AD 35770.AE 35770.AP
Tungsteno 1000 mg/L W in 5% acido nitrico with hydrofluoric acid (max. 1%) (da W) Specpure® Standard monoelemento per ICP
Standard Standard cationici Standard di tungsteno

Molecular Formula: W in 5% HNO₃/tr. HF
Physical Form: Liquid
Odour: Acidic
UN No.: 3264
MDL No.: MFCD00011461


Certificazioni: Tracciabile NIST

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR