Stampa…

Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Bismuth triiodide

Ampuled under argon

35681.06 35681.14
35681.06EA 242 EUR
35681.06 35681.14
Bismuth(III) iodide, ultra dry ≥99.998% (metals basis)
Bismuth(III) iodide

Purity: 99.998% CAS No.: 7787-64-6 Molecular Formula: BiI3 Formula Weight: 589.69 Physical Form: -10 Mesh Granules, Ampouled under argon Density: 5.778 Melting Point: 408° Boiling Point: ca 500° Merck Reference: 14,1269 UN No.: 3260 MDL No.: MFCD00010894

Formula: BiI₃
Punto di ebollizione: ∼500 °C
Punto di fusione: 408 °C
Densità: 5,778
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00010894
CAS Number: 7787-64-6
EINECS: 232-127-4
UN: 3260
ADR: 8,III
Merck Index: 14,1269

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR