Stampa…

Acido cloridrico standardised solution 5.0 N

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Hydrochloric acid solution, Muriatic acid

35638.K2 35638.K7
35638.K2EA 39.7 EUR
35638.K2 35638.K7
Acido cloridrico standardised solution 5.0 N
Acido cloridrico

CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Physical Form: Liquid Appearance: Colourless to pale yellow UN No.: 1789 MDL No.: MFCD00011324

Formula: HCl
Densità: 1,06 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR