Stampa…

Acido solforico standardised solution 5.0 N in soluzione acquosa

Fornitore: Thermo Fisher Scientific
Pericolo

35613.K2
35613.K2EA 45.5 EUR
35613.K2
Acido solforico standardised solution 5.0 N in soluzione acquosa
Acido solforico

CAS No.: 7664-93-9 Molecular Formula: H2SO4 Formula Weight: 98.08 Physical Form: Liquid Appearance: Colourless to pale yellow Odour: Acrid UN No.: 2796 MDL No.: MFCD00064589

Formula: H₂SO₄
Densità: 1,146 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR