Stampa…

Argento nitrato solution_standardised 0.1 N aqueous solution

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

35601.AP 35601.K2 35601.M4
35601.APEA 62.2 EUR
35601.AP 35601.K2 35601.M4
Argento nitrato solution_standardised 0.1 N aqueous solution
Argento nitrato

CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless Odour: Odourless UN No.: 3082 MDL No.: MFCD00003414

Formula: AgNO₃
Peso molecolare: 169.87 g/mol
Punto di ebollizione: 444
Punto di fusione: 210
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR