Stampa…

1-Clorobutano 99,8, CHROMASOLV™ HPLC, Riedel-de Haën™

Fornitore: Honeywell Chemicals
Pericolo

Synonyms: n-Butyl Chloride

34958-2L
34958-2LEA 273.15 EUR
34958-2L
1-Clorobutano 99,8, CHROMASOLV™ HPLC, Riedel-de Haën™
1-Clorobutano

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: CH₃(CH₂)₃Cl
Peso molecolare: 92.57 g/mol
Punto di ebollizione: 78.8 °C (1.013 hPa)
Punto di fusione: –123 °C
Densità: 0.88 g/cm³ (20 °C)
Flash Pt: 10.1 %(V)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001009
CAS Number: 109-69-3
EINECS: 203-696-6
UN: 1127
ADR: 3 II
Merck Index: 13,01559
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2bd980eb6e7ecd91951d46d0ea510119e976dc0&vid=5231549172a609ad96623a80204b11c7c0c69273&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (GC) Min. 99.8 %
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as HCl) Max. 0.001 %
Free alkali (as NH₃) Max. 0.0002 %
Transmittance at 230 nm Min. 65 %
Transmittance at 240 nm Min. 95 %
Transmittance from 250 nm Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR