Stampa…

n-Eptano ≥99% (by GC), CHROMASOLV™ per analisi dei pesticidi, Riedel-de Haën™

Fornitore: Honeywell Chemicals
Pericolo

Synonyms: high purity solvents

34495-7L 34495-1L 34495-2.5L 34495-4X2.5L
34495-7LEA 0 EUR
34495-7L 34495-1L 34495-2.5L 34495-4X2.5L
n-Eptano ≥99% (by GC), CHROMASOLV™ per analisi dei pesticidi, Riedel-de Haën™
n-Eptano

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent

Formula: H₃C(CH₂)₅CH₃
Peso molecolare: 100.20 g/mol
Punto di ebollizione: 98.2…98.4 °C (1.000 hPa)
Punto di fusione: –90.5 °C
Densità: 0.69 g/cm³ (15 °C)
Flash Pt: 6.7 %(V)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (GC) Min. 99 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Suitability for residue analysis Max. 5 ng/l
Autoignition Temperature 433 °F
Refractive Index n20/D 1.387 (lit.)
Determination of P/N ≤5 ng/L
Vapor Pressure 83 mmHg ( 37.7 °C)
Explosion Limit 7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR