Stampa…
Organic reference standard, Deoxynivalenol solution, analytical standard, Supelco®
Fornitore: Merck
34124-2ML
34124-2MLEA 408 EUR
34124-2ML
Organic reference standard, Deoxynivalenol solution, analytical standard, Supelco®
Standard Standard organici
Deoxynivalenol belongs to the group of tetracyclic sesquiterpenoid mycotoxins called trichothecenes, produced by Fusarium fungi, naturally occurring in small grains and infected corn.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2b7afdd856fe1d840baf9e608e00fdcfa42abd6&vid=e2b7afdd856fe1d840baf9e608e00fdcfa42abd6&prid=web&clid=SAPDAM