Stampa…
Organic reference standard, Aflatoxin G1 solution, analytical standard, Supelco®
Fornitore: Merck
34032-2ML-R
34032-2ML-REA 299 EUR
34032-2ML-R
Organic reference standard, Aflatoxin G1 solution, analytical standard, Supelco®
Standard Standard organici
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=63885a786b7574d3c743084ec9950c17e0c88106&vid=63885a786b7574d3c743084ec9950c17e0c88106&prid=web&clid=SAPDAM
Visualizza i codici
Specifiche
Descrizione Aflatoxin G1 solution
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 1165-39-5
Formula C₁₇H₁₂O₇
Temperatura di conservazione –20 °C