Stampa…

Silicic acid hydrate, per analisi

Fornitore: Thermo Fisher Scientific
Attenzione

33270.22 33270.36
33270.22EA 86.4 EUR
33270.22 33270.36
Silicic acid hydrate, per analisi
Silicic acid hydrate

Silicic acid hydrate is used for studying neurodegenerative diseases due to its ability to inhibit aluminum uptake. It is utilized for the manufacture catalyst and catalyst carrier. It is involved in tungsten filament production as chemical reagent and flux. Further, it is used for oil and wax decoloring.


CAS No.: 1343-98-2 Molecular Formula: SiO2.xH2O Formula Weight: 60.09(anhy) Physical Form: -100 Mesh Powder Appearance: White MDL No.: MFCD00054122

Formula: SiO₂·xH₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00054122
CAS Number: 1343-98-2
EINECS: 215-683-2
Merck Index: 14,8490

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR