Stampa…

Dynasore ≥98% (by HPLC), Millipore®

Fornitore: Merck Millipore Calbiochem

324410-10
324410-10EA 158 EUR
324410-10
Dynasore ≥98% (by HPLC), Millipore®
Dynasore

Dynasore, also known as Dynamin inhibitor I, is a cell-permeable semicarbazone compound that inhibits the GTPase activity of dynamin1/2 (IC50 ~15 µM) and Drp1. It has no significant effect against two other small GTPases, MxA and Cdc42.


  • Pale yellow solid
  • Packaged under inert gas
  • Protect from light


Dynasore has been used to elucidate the roles of dynamin in endocytosis mediated by both clathrin-coated and synaptic vesicles. The inhibitition is noncompetitive with respect to GTP and the inhibitor target site presumably resides in the GTPase domain.

Formula: C₁₈H₁₄N₂O₄
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00292551
CAS Number: 304448-55-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR