Stampa…
Campionatore di ghiaccio
Fornitore: BURKLE
5339-1050 5339-0300 5339-0550
300-1049EA 850 EUR
300-1049 300-1047 300-1048
Campionatore di ghiaccio
Campionatori Campionatori per Solidi
Sampler developed for sampling from frozen, highly viscous, paste-like materials such as ice-cream and similar substances. The drill-shaped cutter screws itself into the frozen goods independently when being rotated. The collection cylinder can easily be detached via a bayonet connection. The sample is then available and can be removed.
  • Stainless steel AISI 316L (1.4404), electro-polished
  • Sterilisable
  • Easy cleaning
  • Collection cylinder 38 mm Ø