Stampa…
VWR®, Kit di spugne per campionamento superficiale
Fornitore: VWR Chemicals
710-1020EA 853 EUR
710-1020 300-0229 300-0230 300-0231 300-0232 300-0240
VWR®, Kit di spugne per campionamento superficiale
Tamponi Tamponi per il campionamento
Surface sampling sponges in two forms: Dry sponge sterile sampling bag with write-on strip, containing a dry, biocide-free cellulose sponge, or wet sponge sterile sampling bag, with a foil-sealed, pre-moistened cellulose sponge, and 10 ml of sterile neutralising buffer.
  • Sterilised using gamma irradiation and leakproof transport bags, for the rapid cooling of samples in refrigerators
  • Puncture-proof safety bags eliminating sharp points that can cause puncture and/or abrasions
  • Larger surface in comparison with normal sampling swabs
This surface samplingMore Product Information