Stampa…

Xilene (miscela di isomeri) ≥98.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. per analisi

Fornitore: VWR Chemicals
Attenzione

28975.291EA 87.1 EUR
28975.291 28975.325 28975.360 28975.462 28975.950
Xilene (miscela di isomeri) ≥98.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. per analisi
Xilene (miscela di isomeri)
Formula: C₆H₄(CH₃)₂
Punto di ebollizione: 140 °C (1013 hPa)
Punto di fusione: –34 °C
Densità: 0,86 g/cm³ (20 °C)
Flash Pt: 25 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00077264
CAS Number: 1330-20-7
EINECS: 215-535-7
UN: 1307
ADR: 3,III
REACH: 01-2119488216-32
Merck Index: 13,10136
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2deefc7b7b67c10e64003e243ff3216fad27cf7d&vid=9fa01ddafd4814d5f34266ae7933463a29d92d19&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay (mix of 3 isomers + ethylbenzene) Min. 98.5 %
IR Spectrum Passes test
Acidity or alkalinity Max. 0.0002 meq/g
Boiling point 137 - 141 °C
Colouration Max. 10 APHA
Density (20/4) 0.865 - 0.867
Density (20/20) 0.866 - 0.869
Indice di rifrazione (20°C) 1.495 - 1.499
Substances coloured by H2SO4 Max. 1600 APHA
Benzene Max. 0.02 %
Ethylbenzene Max. 25 %
Evaporation residue Max. 10 ppm
Sulphur compounds (as S) Max. 5 ppm
Thiophene Max. 1 ppm
Toluene Max. 1.0 %
Water Max. 0.03 %
Al (Aluminium) Max. 0.05 ppm
B (Boron) Max. 0.02 ppm
Ba (Barium) Max. 0.05 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.02 ppm
Cu (Copper) Max. 0.02 ppm
Fe (Iron) Max. 0.05 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.05 ppm
Ni (Nickel) Max. 0.01 ppm
Pb (Lead) Max. 0.02 ppm
Sn (Tin) Max. 0.05 ppm
Zn (Zinc) Max. 0.05 ppm
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR