Stampa…

Titanio (III) cloruro 15% in hydrochloric acid 10%, AnalaR NORMAPUR® per analisi

Fornitore: VWR Chemicals
Pericolo

Synonyms: Titanium trichloride

28660.236EA 75.5 EUR
28660.236 28660.293
Titanio (III) cloruro 15% in hydrochloric acid 10%, AnalaR NORMAPUR® per analisi
Titanio (III) cloruro
Formula: Cl₃Ti
Densità: 1,2 g/cm³ (20 °C)
MDL Number: MFCD00011266
CAS Number: 7705-07-9
EINECS: 231-728-9
UN: 1789
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Assay 12.5 a 17.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR