Stampa…

Setaccio molecolare 3A (0,3 nm, 3 Å), in astine, GPR RECTAPUR®, Ø ~1.6 mm

Fornitore: VWR Chemicals

28461.292EA 250 EUR
28461.292 28461.230
Setaccio molecolare 3A (0,3 nm, 3 Å), in astine, GPR RECTAPUR®, Ø ~1.6 mm
Setaccio molecolare 3A (0,3 nm, 3 Å)

This molecular sieve is a clay-bound potassium form of Type A molecular sieve. A 3Å molecular sieve will adsorb molecules with critical diameters up to three angstroms. It adsorbs water and ammonia, but not larger molecules like ethane.


  • Equilibrium H2O capacity: ~ 21% (wt) (measured at 17.5mm Hg; 25°C)

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00131613
CAS Number: 308080-99-1
EINECS: 215-283-8

Visualizza i codici

Risultati del test di specificazione

Identification Passes test
Loss on ignition (575 °C) max. 2.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR