Stampa…
Kit di selezione per colonne cromatografiche a interazione idrofobica, HiTrap™ HIC
Fornitore: Cytiva
28-4110-07
28-4110-07EA 321 EUR
28-4110-07
Kit di selezione per colonne cromatografiche a interazione idrofobica, HiTrap™ HIC
Colonne cromatografiche
HiTrap HIC selection kit is designed for rapid screening for the most appropriate hydrophobic interaction chromatography (HIC) resin and experimental conditions.
  • High likelihood of finding suitable HIC resin – contains seven different resins
  • Convenient prepacked HiTrap columns, compatible with syringe, single laboratory pump, or chromatography systems such as ÄKTA systems
Within HIC you need to perform empirical testing to find optimal conditions for your target sample. The parameters you typically screen for are type of salt, concentration of salt,More Product Information