Stampa…

Sodio idrogeno carbonato 99.0-101.0% Ph. Eur.

Fornitore: VWR Chemicals

27775.293EA 67.8 EUR
27775.293 27775.362 27775.464
Sodio idrogeno carbonato 99.0-101.0% Ph. Eur.
Sodio idrogeno carbonato
Formula: NaHCO₃
Punto di fusione: 270 °C
Densità: 2,159 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Assay 99.0 - 101.0 %
Appearance: .
White or almost white crystalline powder Passes test
Identification A Passes test
Identification B Passes test
Identification (Na) Passes test
Appearance of solution Passes test
CO3 (Carbonate) Passes test
Cl (Chloride) ≤ 150 ppm
SO4 (Sulphate) ≤ 150 ppm
NH4 (Ammonium) ≤ 20 ppm
Ca (Calcium) ≤ 100 ppm
Fe (Iron) ≤ 20 ppm
Heavy metals (as Pb) ≤ 10 ppm
Residual solvents Unlikely by manuf.process
As (Arsenic) ≤ 2 ppm
Cd (Cadmium) ≤ 1 ppm
Co (Cobalt) ≤ 1 ppm
Hg (Mercury) ≤ 2 ppm
Ni (Nickel) ≤ 1 ppm
Pb (Lead) ≤ 3 ppm
V (Vanadium) ≤ 1 ppm
Conforms to Ph.Eur. Passes test
Original manufacturer code R or B


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR