Stampa…

D(+)-Saccarosio, AnalaR NORMAPUR® per analisi

Fornitore: VWR Chemicals

27480.294EA 81 EUR
27480.294 27480.260 27480.360
D(+)-Saccarosio, AnalaR NORMAPUR® per analisi
D(+)-Saccarosio
Formula: C₁₂H₂₂O₁₁
Punto di fusione: 169…170 °C
Densità: 1,29 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Acidity Max. 0.005 meq/g
Specific optical rotation (26 %; water) 66.4 - 66.8 °
Glucose Max. 0.05 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 80 ppm
Insolubility in water Max. 30 ppm
Loss on drying (100°C) Max. 0.05 %
Reducing sugars (calculated as C6H12O6) Max. 0.1 %
Cl (Chloride) Max. 5 ppm
N (Nitrogen) Max. 20 ppm
SO3 (as SO2) Max. 15 ppm
SO4 (Sulphate) Max. 20 ppm
Ba (Barium) Max. 0.02 %
Ca (Calcium) Max. 10 ppm
Cd (Cadmium) Max. 0.05 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.5 ppm
Ni (Nickel) Max. 0.1 ppm
Pb (Lead) Max. 0.5 ppm
Zn (Zinc) Max. 0.5 ppm

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR