Stampa…

D(+)-Saccarosio, GPR RECTAPUR®

Fornitore: VWR Chemicals

Synonyms: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose

27478.296EA 71.4 EUR
27478.296 27478.365 27478.467
D(+)-Saccarosio, GPR RECTAPUR®
D(+)-Saccarosio
Formula: C₁₂H₂₂O₁₁
Punto di fusione: 169…170 °C
Densità: 1,29 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Specific optical rotation (2 %; water) 66 - 67 °
Heavy metals (as Pb) Max. 50 ppm
Ignition residue (SO4) Max. 0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR