Stampa…
VWR® Talon®, Clamps, Fixed Position, Two Pronged
Fornitore: VWR Collection
241-0409EA 131 EUR
241-0409
VWR® Talon®, Clamps, Fixed Position, Two Pronged
Morsetti Clamp per uso generico
Fixed-position clamps made of stainless steel with electro-polished finish. Used to hold apparatus near the lab frame where no adjustment is required after set up.
  • With non-slip vinyl sleeves
  • Built-in holder grips rods up to 19 mm Ø
  • Can be rotated after attachment to a lab frame or other apparatus
Informazioni per l'ordine: Supplied with non-slip vinyl sleeves, and with fiberglass covers for temperatures above 100 °C (212 °F).