Stampa…

Anti-COX2, C-Terminal Rabbit Polyclonal Antibody

Fornitore: Merck Millipore (Oncogene)
236002-50
236002-50EA 716 EUR
236002-50
Anti-COX2, C-Terminal Rabbit Polyclonal Antibody
Anticorpi
Type: Primary
Antigen: COX2, C-Terminal
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR