Stampa…
Ball Valve for Storage Bottles
Fornitore: SEMADENI
3100
229-7489EA 107 EUR
229-7489
Ball Valve for Storage Bottles
Valvole Valvola a sfera
½” universal valve made from PP, resistant to acids, alkalis and solvents.
  • Fully sealed design, runs very easily and with no blockages
  • Outlet spout Ø 12 mm
Housing and ball made from PP, ball seal made from PTFE and O ring made from nitrile rubber (NBR 70).